Anti-YES1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Lieferant: United States Biological
USBIY2058-PEEA 852 EUR
USBIY2058-PE
Anti-YES1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: YES1 (v-yes-1 Yamaguchi sarcoma viral oncogene homolog 1)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR