Anti-YY1AP1 Rabbit Polyclonal Antibody

Lieferant: US Biological
Y1200
USBIY1200EA 554 EUR
USBIY1200
Anti-YY1AP1 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Flow Cytometry: 1:50; Western Blot: 1:1000; Immunoprecipitation: 1:50; Immunochemistry:1:25

Type: Primary
Antigen: YY1AP1 (YY1 associated protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR