Anti-XRCC6 Mouse Monoclonal Antibody [clone: 11C966]

Lieferant: US Biological
X9506-02
USBIX9506-02EA 522 EUR
USBIX9506-02
Anti-XRCC6 Mouse Monoclonal Antibody [clone: 11C966]
Antikörper
Recommended dilution: ELISA: 1 ?g/ml-3ng/ml; Western Blot: 1:500-1:1000; Immunochemistry: 1. 5 ?g/ml

Type: Primary
Antigen: XRCC6 (X-ray repair complementing defective repair in Chinese hamster cells 6)
Clonality: monoclonal
Clone: 11C966
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR