Anti-XRCC5 Mouse Monoclonal Antibody

Lieferant: US Biological
X9505-13D
USBIX9505-13DEA 390 EUR
USBIX9505-13D
Anti-XRCC5 Mouse Monoclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1 ?g/ml; Immunofluorescence: 1-4?g/ml

Type: Primary
Antigen: XRCC5 (X-ray repair complementing defective repair in Chinese hamster cells 5 (double-strand-break rejoining))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR