Anti-ERCC5 Mouse Monoclonal Antibody [clone: 8J112]

Lieferant: United States Biological
USBIX1048-11BEA 574 EUR
USBIX1048-11B
Anti-ERCC5 Mouse Monoclonal Antibody [clone: 8J112]
Antikörper
Type: Primary
Antigen: ERCC5 (excision repair cross-complementation group 5)
Clonality: monoclonal
Clone: 8J112
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR