Anti-XK Rabbit Polyclonal Antibody

Lieferant: US Biological
X1037-01A
USBIX1037-01AEA 633 EUR
USBIX1037-01A
Anti-XK Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: XK (X-linked Kx blood group (McLeod syndrome))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR