Anti-XIAP Rabbit Polyclonal Antibody

Lieferant: US Biological
X1034-10G
USBIX1034-10GEA 461 EUR
USBIX1034-10G
Anti-XIAP Rabbit Polyclonal Antibody
Antikörper
  • Recommended dilution: Western Blot: 0.5-4μg/ml

Type: Primary
Antigen: XIAP (X-linked inhibitor of apoptosis)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR