Anti-XIAP Rabbit Polyclonal Antibody

Lieferant: US Biological
X1033-98E
USBIX1033-98EEA 437 EUR
USBIX1033-98E
Anti-XIAP Rabbit Polyclonal Antibody
Antikörper
  • Recommended dilution: Western Blot: 1:1000-1:2000; Immunochemistry: 1:1000-1:5000; Immunoprecipitation: 1:50-1:200

Type: Primary
Antigen: XIAP (X-linked inhibitor of apoptosis)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR