Anti-EDA2R Mouse Monoclonal Antibody [clone: 11C964]

Lieferant: US Biological
X1011-51A
USBIX1011-51AEA 522 EUR
USBIX1011-51A
Anti-EDA2R Mouse Monoclonal Antibody [clone: 11C964]
Antikörper
Recommended dilution: Western Blot: 1:500-1:1000; ELISA: 1 ?g/ml-3ng/ml

Type: Primary
Antigen: EDA2R (ectodysplasin A2 receptor)
Clonality: monoclonal
Clone: 11C964
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR