Anti-XBP1 Chicken Polyclonal Antibody

Lieferant: United States Biological
USBIX0989-52CEA 559 EUR
USBIX0989-52C
Anti-XBP1 Chicken Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:2000; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: XBP1 (X-box binding protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR