Anti-WIF1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Lieferant: US Biological
W3005-75B-APC
USBIW3005-75B-APCEA 852 EUR
USBIW3005-75B-APC
Anti-WIF1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:50-1:100; Immunochemistry: 1:10-1:50
Do not freeze!

Type: Primary
Antigen: WIF1 (WNT inhibitory factor 1)
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR