Anti-WNK4 Rabbit Polyclonal Antibody

Lieferant: US Biological
W2999-05
USBIW2999-05EA 587 EUR
USBIW2999-05
Anti-WNK4 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: WNK4 (WNK lysine deficient protein kinase 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR