Anti-WNK3 Rabbit Polyclonal Antibody (Biotin)

Lieferant: United States Biological
USBIW2998-04-BIOTEA 852 EUR
USBIW2998-04-BIOT
Anti-WNK3 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: WNK3 (WNK lysine deficient protein kinase 3)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR