Anti-WNK1 Rabbit Polyclonal Antibody (Biotin)

Lieferant: United States Biological
USBIW2996-01C-BIOTEA 852 EUR
USBIW2996-01C-BIOT
Anti-WNK1 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: WNK1 (WNK lysine deficient protein kinase 1)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR