Anti-AMER1 Rabbit Monoclonal Antibody [clone: D38E5]

Lieferant: US Biological
W1125-18A
USBIW1125-18AEA 554 EUR
USBIW1125-18A
Anti-AMER1 Rabbit Monoclonal Antibody [clone: D38E5]
Antikörper
Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: AMER1 (APC membrane recruitment protein 1)
Clonality: monoclonal
Clone: D38E5
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR