Anti-WEE1 Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
W1015-01C-BIOT
USBIW1015-01C-BIOTEA 852 EUR
USBIW1015-01C-BIOT
Anti-WEE1 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: WEE1 (WEE1 G2 checkpoint kinase)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR