Anti-WASF3 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIW0952-01EA 515 EUR
USBIW0952-01
Anti-WASF3 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:1000; Immunoprecipitation: 1:50

Type: Primary
Antigen: WASF3 (WAS protein family, member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Monkey, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR