Anti-VWF Mouse Monoclonal Antibody [clone: 2Q2134]

Lieferant: United States Biological
USBIV2700-08AEA 681 EUR
USBIV2700-08A
Anti-VWF Mouse Monoclonal Antibody [clone: 2Q2134]
Antikörper
Type: Primary
Antigen: VWF (von Willebrand factor)
Clonality: monoclonal
Clone: 2Q2134
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR