Anti-VTN Rabbit Polyclonal Antibody

Lieferant: US Biological
V2200-02G
USBIV2200-02GEA 823 EUR
USBIV2200-02G
Anti-VTN Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Immunochemistry: 1:50

Type: Primary
Antigen: VTN (vitronectin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR