Anti-Vitellogenin Mouse Monoclonal Antibody [clone: 6D844]

Lieferant: US Biological
V2150-59D
USBIV2150-59DEA 529 EUR
USBIV2150-59D
Anti-Vitellogenin Mouse Monoclonal Antibody [clone: 6D844]
Antikörper
Recommended dilution: ELISA: 1:100-1:1000

Type: Primary
Antigen: Vitellogenin
Clonality: monoclonal
Clone: 6D844
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Zebrafish
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR