Anti-MAVS Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIV2124-20EA 526 EUR
USBIV2124-20
Anti-MAVS Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: MAVS (mitochondrial antiviral signaling protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR