Anti-VIM Rabbit Polyclonal Antibody (FITC (Fluorescein))

Lieferant: US Biological
V2122-10P-FITC
USBIV2122-10P-FITCEA 852 EUR
USBIV2122-10P-FITC
Anti-VIM Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antikörper
Recommended dilution: Flow Cytometry: 1:10-1:50; Immunochemistry: 1:10-1:50; Western Blot: 1:1000
Do not freeze!

Type: Primary
Antigen: VIM (vimentin)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR