Anti-VIM Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Lieferant: United States Biological
USBIV2122-10P-APCEA 791 EUR
USBIV2122-10P-APC
Anti-VIM Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: Flow Cytometry: 1:10-1:50; Immunochemistry: 1:10-1:50; Western Blot: 1:1000
Do not freeze!

Type: Primary
Antigen: VIM (vimentin)
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR