Anti-VIM Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIV2122-01GEA 343 EUR
USBIV2122-01G
Anti-VIM Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 0. 05-0.2?g/ml; Western Blot: 0.5-1 ?g/ml; Immunochemistry: 5-10 ?g/ml

Type: Primary
Antigen: VIM (vimentin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR