Anti-SLC18A2 Rabbit Polyclonal Antibody

Lieferant: US Biological
V2121-89D
USBIV2121-89DEA 776 EUR
USBIV2121-89D
Anti-SLC18A2 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Immunofluorescence: 1:300; Immunochemistry: 1:2000-4000

Type: Primary
Antigen: SLC18A2 (solute carrier family 18 (vesicular monoamine transporter), member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR