Anti-VEGFC Rabbit Polyclonal Antibody

Lieferant: US Biological
V2121-30A
USBIV2121-30AEA 695 EUR
USBIV2121-30A
Anti-VEGFC Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 0.1-1 ?g/ml; Western Blot: 2-5 ?g/ml

Type: Primary
Antigen: VEGFC (vascular endothelial growth factor C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR