Anti-VAV3 Rabbit Polyclonal Antibody

Lieferant: US Biological
V2121-15B
USBIV2121-15BEA 544 EUR
USBIV2121-15B
Anti-VAV3 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Immunoblot: 0.5-2?g/ml

Type: Primary
Antigen: VAV3 (vav 3 guanine nucleotide exchange factor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR