Anti-Vasostatin I Rabbit Polyclonal Antibody

Lieferant: US Biological
V2120-44D
USBIV2120-44DEA 979 EUR
USBIV2120-44D
Anti-Vasostatin I Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:12800

Type: Primary
Antigen: Vasostatin I
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR