Anti-SERPINA12 Sheep Polyclonal Antibody (Biotin)

Lieferant: United States Biological
USBIV2118-10BEA 795 EUR
USBIV2118-10B
Anti-SERPINA12 Sheep Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: Western Blot: 0.1-0.2?g/ml

Type: Primary
Antigen: SERPINA12 (serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 12)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR