Anti-VASP Mouse Monoclonal Antibody [clone: 9G459]

Lieferant: United States Biological
USBIV2118-01KEA 601 EUR
USBIV2118-01K
Anti-VASP Mouse Monoclonal Antibody [clone: 9G459]
Antikörper
Type: Primary
Antigen: VASP (vasodilator-stimulated phosphoprotein)
Clonality: monoclonal
Clone: 9G459
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR