Anti-VIP Rabbit Polyclonal Antibody

Lieferant: US Biological
V2115-11A
USBIV2115-11AEA 736 EUR
USBIV2115-11A
Anti-VIP Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: VIP (vasoactive intestinal peptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Pig, Rat, Sheep
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR