Anti-VIP Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIV2115-06CEA 530 EUR
USBIV2115-06C
Anti-VIP Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Immunoprecipitation: 1:500

Type: Primary
Antigen: VIP (vasoactive intestinal peptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR