Anti-FLT1 Mouse monoclonal antibody [clone: 9F651]

Lieferant: US Biological
V2110-16D
USBIV2110-16DEA 537 EUR
USBIV2110-16D
Anti-FLT1 Mouse monoclonal antibody [clone: 9F651]
Antikörper
Recommended dilution: ELISA: 1:8000

Type: Primary
Antigen: FLT1 (fms-related tyrosine kinase 1)
Clonality: monoclonal
Clone: 9F651
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR