Anti-VAPB Sheep Polyclonal Antibody

Lieferant: United States Biological
USBIV2024-15AEA 793 EUR
USBIV2024-15A
Anti-VAPB Sheep Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1 ?g/ml; ELISA: 0.5-1 ?g/ml

Type: Primary
Antigen: VAPB (VAMP (vesicle-associated membrane protein)-associated protein B and C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR