Anti-USP33 Mouse Monoclonal Antibody [clone: 5B5]

Lieferant: United States Biological
USBIU4101-36AEA 703 EUR
USBIU4101-36A
Anti-USP33 Mouse Monoclonal Antibody [clone: 5B5]
Antikörper
Recommended dilution: Western Blot: 1:500-1:1000; Immunofluorescence: 20 ?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: USP33 (ubiquitin specific peptidase 33)
Clonality: monoclonal
Clone: 5B5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR