Anti-USP28 Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
U4101-28D-BIOT
USBIU4101-28D-BIOTEA 852 EUR
USBIU4101-28D-BIOT
Anti-USP28 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: USP28 (ubiquitin specific peptidase 28)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR