Anti-USP25 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Lieferant: US Biological
U4101-25B-APC
USBIU4101-25B-APCEA 852 EUR
USBIU4101-25B-APC
Anti-USP25 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: USP25 (ubiquitin specific peptidase 25)
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR