Anti-USP21 Goat Polyclonal Antibody

Lieferant: US Biological
U4101-06H
USBIU4101-06HEA 420 EUR
USBIU4101-06H
Anti-USP21 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:64000

Type: Primary
Antigen: USP21 (ubiquitin specific peptidase 21)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR