Anti-USP21 Rabbit Polyclonal Antibody

Lieferant: US Biological
U4101-06C
USBIU4101-06CEA 587 EUR
USBIU4101-06C
Anti-USP21 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: USP21 (ubiquitin specific peptidase 21)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR