Anti-USP11 Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
U4100-20B-BIOT
USBIU4100-20B-BIOTEA 852 EUR
USBIU4100-20B-BIOT
Anti-USP11 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: USP11 (ubiquitin specific peptidase 11)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR