Anti-USP7 Rabbit Polyclonal Antibody

Lieferant: US Biological
U4100-02D
USBIU4100-02DEA 587 EUR
USBIU4100-02D
Anti-USP7 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: USP7 (ubiquitin specific peptidase 7 (herpes virus-associated))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR