Anti-USP4 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIU4099-86GEA 704 EUR
USBIU4099-86G
Anti-USP4 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Immunochemistry: 5-10 ?g/ml

Type: Primary
Antigen: USP4 (ubiquitin specific peptidase 4 (proto-oncogene))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR