Anti-USP4 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Lieferant: US Biological
U4099-85B-FITC
USBIU4099-85B-FITCEA 852 EUR
USBIU4099-85B-FITC
Anti-USP4 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antikörper
Recommended dilution: FLISA: 1:1000; Immunochemistry: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: USP4 (ubiquitin specific peptidase 4 (proto-oncogene))
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR