Anti-USP3 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Lieferant: US Biological
U4099-70B-FITC
USBIU4099-70B-FITCEA 852 EUR
USBIU4099-70B-FITC
Anti-USP3 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antikörper
Recommended dilution: FLISA: 1:1000; Immunochemistry: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: USP3 (ubiquitin specific peptidase 3)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR