Anti-USP2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Lieferant: US Biological
U4099-60C-PE
USBIU4099-60C-PEEA 852 EUR
USBIU4099-60C-PE
Anti-USP2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!

Type: Primary
Antigen: USP2 (ubiquitin specific peptidase 2)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR