Anti-UCN3 Rabbit Polyclonal Antibody

Lieferant: US Biological
U2575-33A
USBIU2575-33AEA 979 EUR
USBIU2575-33A
Anti-UCN3 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:12800; Immunochemistry: 1:200

Type: Primary
Antigen: UCN3 (urocortin 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR