Anti-UCK2 Rabbit Polyclonal Antibody

Lieferant: US Biological
U2281-03
USBIU2281-03EA 587 EUR
USBIU2281-03
Anti-UCK2 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:250; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: UCK2 (uridine-cytidine kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR