Anti-UCK2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Lieferant: United States Biological
USBIU2281-02-APCEA 852 EUR
USBIU2281-02-APC
Anti-UCK2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Immunochemistry: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: UCK2 (uridine-cytidine kinase 2)
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR