Anti-ULK1 Rabbit Polyclonal Antibody

Lieferant: US Biological
U1500-70E
USBIU1500-70EEA 507 EUR
USBIU1500-70E
Anti-ULK1 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1-3?g/ml; Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: ULK1 (unc-51 like autophagy activating kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR