Anti-TRIB3 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIT8335-43HEA 507 EUR
USBIT8335-43H
Anti-TRIB3 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1-3?g/ml

Type: Primary
Antigen: TRIB3 (tribbles pseudokinase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR